      • Stem Cell Research
      PCR with higher success, simpler success, and a trusted enzyme
      1:46
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      Thermo Scientific™ Phusion™ High‑Fidelity PCR enzyme has been upgraded. The latest version, Phusion™ Plus DNA Polymerase, features: • A buffer formulated for a universal annealing temperature—no more calculating primer annealing temperatures • Increased enzyme fidelity of >100x that of Taq DNA polymerase—fewer PCR sequence errors • More robust performance with challenging templates like GC‑rich sequences, low‑abundance targets, and DNA of suboptimal purity Learn more at https://thermofisher.com/phusionplus
      View More
      View Less
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels