      • Stem Cell Research
      How to set up an Invitrogen QuantiGene Plex Assay in the Luminex xPONENT Software
      8:11
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      "In this video we will demonstrate how to set up an Invitrogen™ QuantiGene Plex Assay in the Luminex® xPONENT™ Software . We will walk you through the following procedures: 1) Log in and perform system initialization 2) Create a new protocol 3) Add new standard lots 4) Create a new batch 5) Read plate/Run batch 6) Evaluate and export results for further analysis on the Thermo Fisher Scientific ProcartaPlex Analysis App (https://apps.thermofisher.com/apps/quantigene/) For more information, please refer to the Invitrogen™ QuantiGene™ Plex user guide or contact Technical Support. Invitrogen™ QuantiGene™ Assays: https://www.thermofisher.com/qgp Invitrogen™ QuantiGene™ Analysis App: https://apps.thermofisher.com/apps/quantigene/ If you have any questions or concerns about running Invitrogen™ QuantiGene™ Plex assay, please contact Thermo Fisher Scientific Technical Service: eutotech@thermofisher.com (Europe) or techservices@thermofisher.com (North America). Or visit our Support Centers: https://www.thermofisher.com/antibodysupport"
      View More
      View Less
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels