      • Stem Cell Research
      Invitrogen Bigfoot Cell Sorter - Overview
      3:43
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      Sorting real-time, spectrally unmixed samples allows higher parameter panels. The experiment wizard assists the user in running controls, identifying potential issues, and applying the unmixing algorithms to produce high-quality data. The Bigfoot Spectral Cell Sorter can be configured with up to 9 lasers and 60 detectors, providing the versatility for both compensation and spectral unmixing. The multiple scatter options offer high flexibility with simultaneous standard, small particle, multi-laser scatter detection, and polarization options. By utilizing the massively parallel, pipelined architecture, the system allows complex, high parameter experiments with up to 60 x 60 compensation or spectral unmixing without limiting instrument performance.
      View More
      View Less
      Learn more about the Invitrogen Bigfoot Cell Sorter
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels