Invitrogen Bigfoot Cell Sorter - Spectral Unmixing
1:41
Description
Related Videos
The Invitrogen Bigfoot Spectral Cell Sorter utilizes custom electronics specifically designed for cell sorting and is capable of sorting on spectrally unmixed data in real time. The optional spectral mode enables higher parameter experiments than are typically done with conventional flow cytometry. The experiment wizard assists with running controls, identifying potential issues, and applying the unmixing algorithms to produce high-quality data.