Webinar: Identification of protein aggregation through UV-Visible spectroscopy: The use of absorption and diffuse reflection techniques
44:41
Protein aggregates can irreversibly form in solution through a number of processes and are common within unstable samples. The resulting lower concentration of non-aggregated materials can negatively impact pharmaceutical applications and can require further purification steps. Consequently, it is important to determine the extent to which a sample is aggregated early in the experiment. Typically, aggregates are identified using size exclusion chromatography. However, recovering the samples after this analysis is difficult and often time consuming. In this webinar, Dr. Jennifer Empey, Thermo Fisher Scientific, demonstrates how to rapidly recognize the presence of protein aggregates using UV-Visible absorption spectroscopy, a non-destructive analysis technique.