Save time by combining two workflows with the Attune(TM) CytPix Flow Cytometer
1:23
Description
Related Videos
The Invitrogen™ Attune™ CytPix Flow Cytometer can analyze quantitative image parameter data concurrently with multiparameter flow cytometry data. No need to split your sample or modify your flow cytometry protocols. Simplify your imaging and flow cytometry workflows and save time. Learn more at thermofisher.com/cytpix