Fast, Flexible, and Spectrally Brilliant: Invitrogen Attune Xenith Flow Cytometer
1:00
Fast, flexible, and spectrally brilliant—simplify high-speed, high-parameter analysis with the Invitrogen Attune Xenith Flow Cytometer, powered by our legacy core acoustic hydrodynamic focusing technology. Experience remarkable speed, operating five times faster than most spectral cytometers, and benefit from innovative flexibility with seamless transitions between spectral unmixing and conventional flow cytometry on the same instrument using the same sample. Designed to handle challenging samples without clogging, the Attune Xenith Flow Cytometer boasts an advanced optical design featuring 6 lasers, 51 fluorescence detectors, and 6 scatter channels. Enjoy simplified maintenance with an intuitive touchscreen and advanced remote access diagnostics, including onboard cameras to ensure uninterrupted performance. Learn how the Invitrogen Attune Xenith Flow Cytometer can transform your research with speed, flexibility, and unparalleled performance. Visit thermofisher.com/attunexenith for more information.