      • Stem Cell Research
      Bitumen and asphalt testing - Dynamic Shear Rheometer (DSR) Thermo Scientific HAAKE MARS iQ Air Rheometer
      4:12
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      The Thermo Scientific HAAKE MARS iQ Rheometer for dynamic shear testing according to AASHTO, ASTM, FGSV and DIN EN for bitumen and asphalt is ideal for routine test laboratories. The rheometer employs several smart functionalities to ensure precise and consistent results. Depending on the standard, it can be equipped with different measuring geometries and an appropriate temperature control. Changing between different configurations is easy, fast and failure-free with “Connect Assist” technology. You can initiate company specific SOPs via the convenient instrument touchscreen or with the rheometer software on the PC and always ensure, that the correct protocol is followed during the entire measurement. Some of the QC features of the HAAKE MARS iQ Rheometers and the HAAKE RheoWin Software are: 1. User guidance, with embedded individual videos and pictures to prevent handling failures 2. Automated data evaluation using your QC criteria to ensure correct result interpretation 3. Generation of reports according to specific company standards with automated data storage 4. Software offers real time display of graphical, numerical measuring data. Via touchscreen numerical real time data & measuring range bars can be monitored.
      View More
      View Less
      Learn More about HAAKE MARS iQ Rheometers for Intuitive, reliable and flexible rheological measurements
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Related Videos
      In Manufacturing & Processing Videos
      • Play video Accelerate your assay development with Custom Commercial Supply by Thermo Fisher Scientific
        Accelerate your assay development with Custom Commercial Supply by Thermo Fisher Scientific
        Custom Commercial Supply can support your assay development with custom solutions, secure supply chain, diversity of experience, & quality raw materials.
        0:45
      • Play video Advantages of continuous electrode slurry compounding with twin-screw extrusion
        Advantages of continuous electrode slurry compounding with twin-screw extrusion
        See how continuous twin-screw mixing assures a constant, high dispersion quality and has a higher yield compared to conventional mixers. Twin-screw extrusion is fast, scalable, and is easy to clean versus planetary mixers.
        2:28
      • Play video Thermo Scientific Niton XL5e Handheld Analyzers
        Thermo Scientific Niton XL5e Handheld Analyzers
        Meet the demands of your work with powerful detection capabilities, excellent ergonomics, and Bluetooth and Wi-Fi connectivity.
        0:38
      • Play video Accelerate your assay development with Custom Commercial Supply by Thermo Fisher Scientific
        Accelerate your assay development with Custom Commercial Supply by Thermo Fisher Scientific
        Custom Commercial Supply can support your assay development with custom solutions, secure supply chain, diversity of experience, & quality raw materials.
        1:42
      • Play video Freeze it and forget it: the value of having a robust formulation and cryopreservation strategy for cell therapy manufacturing
        Freeze it and forget it: the value of having a robust formulation and cryopreservation strategy for cell therapy manufacturing
        Listen to expert Rupa Pike discuss the value of having a robust formulation and cryopreservation strategy for cell therapy manufacturing
        4:05
      • Play video Accelerate your assay development with Custom Commercial Supply by Thermo Fisher Scientific
        Accelerate your assay development with Custom Commercial Supply by Thermo Fisher Scientific
        Custom Commercial Supply can support your assay development with custom solutions, secure supply chain, diversity of experience, & quality raw materials.
        0:30
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels