      • Stem Cell Research
      DNA polymerase properties for PCR
      2:24
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      The key to PCR is the DNA polymerase that synthesizes new DNA strands. General characteristics of DNA polymerases include: • Thermostability • Processivity • Speed • Fidelity, and • Specificity Some DNA polymerases can withstand high temperatures longer than others over the course of PCR cycling. This is often described as the enzyme’s thermostability or half-life at a specific temperature. All DNA polymerases have 5-prime to 3-prime polymerase activity to synthesize new DNA strands. The number of nucleotides the DNA polymerase can incorporate without dissociating from the template defines its processivity. Processivity generally depends on the enzyme’s affinity for the DNA template during amplification. High processivity often contributes to the DNA polymerase’s speed, as well as resistance to inhibitors. The higher the processivity of a DNA polymerase, the faster it can synthesize the new strands. Fast enzymes are especially beneficial in amplifying long templates. Some DNA polymerases also have 3-prime to 5-prime exonuclease activity to remove mismatched nucleotides. This is also called proofreading activity. Proofreading DNA polymerases are used for high-fidelity PCR because they make fewer mistakes during DNA amplification. DNA polymerases without 3-prime to 5-prime exonuclease activity are known as non-proofreading or low-fidelity PCR enzymes. They are mainly used for standard PCR. One common issue encountered in PCR is low specificity or background amplification. PCR specificity can be improved by modifying DNA polymerases to be inactive at room temperature during reaction setup, but activated at a higher temperature at the start of PCR cycling. This approach is commonly known as hot-start PCR.
      View More
      View Less
      Learn more about PCR
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Related Videos
      In PCR Videos
      • Play video Absolute Gene-ius: S2 E8 Now that’s what I call a high-quality viral vector
        Absolute Gene-ius: S2 E8 Now that’s what I call a high-quality viral vector
        Dr. Cliff Froelich covers what vectors his team works with, what methods and technologies they use to monitor quality and safety, and he reminds us why it all matters.
        32:29
      • Play video Love of dPCR - Fundamentals
        Love of dPCR - Fundamentals
        Webinar covering the principles of dPCR and its uses.
        47:42
      • Play video resDNASEQ Cinematic
        resDNASEQ Cinematic
        Video the features of the resDNASEQ workflow
        0:26
      • Play video ProFlex PCR Cinematic
        ProFlex PCR Cinematic
        Increase productivity with our PCR workflow solutions.
        0:34
      • Play video Absolute Gene-ius: S2 E11 Somatic mosaic loss of sex chromosomes and kidney disease
        Absolute Gene-ius: S2 E11 Somatic mosaic loss of sex chromosomes and kidney disease
        Join us to hear about Dr. Parker Wilson's exciting work where he uses digital PCR and NGS methods to identify and quantify rare mutations associated with kidney disease.
        34:08
      • Play video Going Green with PCR to Detecting Grapevine Pathogens, with Lexi Heger| Absolute Gene-ius Podcast S4E1
        Going Green with PCR to Detecting Grapevine Pathogens, with Lexi Heger| Absolute Gene-ius Podcast S4E1
        In this episode of Absolute Gene-ius, Lexi Heger, PhD candidate at Michigan State University, explores how qPCR and digital PCR are transforming plant pathology.
        36:05
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels