      • Stem Cell Research
      Benefits of a universal annealing temperature for PCR
      2:40
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      Primer annealing is a critical step in the polymerase chain reaction, or PCR. Suboptimal annealing can drastically reduce the yield and specificity of your PCR, and even cause it to fail. When it's not possible to design primers with similar melting temperatures, or Tm, the primer with the higher Tm could bind to unintended targets, while the primer with the lower Tm would have difficulty binding at an annealing temperature chosen for these primers. This can cause your PCR to fail. To mitigate this issue, the annealing temperature of each primer set should be optimized. But this can be a long and tedious process. To address the need for optimization, Thermo Fisher Scientific has developed a PCR buffer with isostabilizing components, which increase the stability of primer-template duplexes during annealing. Using PCR enzymes with this buffer, successful amplification can be achieved at 60 degrees Celsius, even with primers of different melting temperatures. Another benefit is that the enhanced stability of primer-template duplexes in this innovative buffer allows for a single PCR extension time with amplicons of different lengths. Typically, a long extension time would result in nonspecific amplification, but the isostabilizing components help achieve PCR specificity, even with a longer extension time. This universal annealing buffer has been developed with Invitrogen Platinum II Taq DNA polymerase, and Platinum SuperFi II DNA polymerase, and Platinum Direct PCR Universal Master Mix. One of a kind, this buffer enables high yield and specificity at 60 degrees Celsius, while also allowing for PCR of short and long amplicons at the same time and using the same protocol, instead of sequential runs. Only offered by Thermo Fisher Scientific, the universal annealing buffer of the Platinum DNA Polymerases helps save time and simplify PCR protocols by: - Reducing the need for optimization of primer annealing, - Enabling primer-template annealing at 60 degrees Celsius, and - Allowing co-cycling of different PCR targets To learn more about these PCR benefits, please visit https://thermofisher.com/pcrenzymes
      View More
      View Less
      Invitrogen Platinum PCR products with universal annealing buffer
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Related Videos
      In PCR Videos
      • Play video Love of dPCR - Fundamentals
        Love of dPCR - Fundamentals
        Webinar covering the principles of dPCR and its uses.
        47:42
      • Play video Absolute Gene-ius: S2 E8 Now that’s what I call a high-quality viral vector
        Absolute Gene-ius: S2 E8 Now that’s what I call a high-quality viral vector
        Dr. Cliff Froelich covers what vectors his team works with, what methods and technologies they use to monitor quality and safety, and he reminds us why it all matters.
        32:29
      • Play video Absolute Gene-ius: S2 E10 Automating accuracy – an insider’s view
        Absolute Gene-ius: S2 E10 Automating accuracy – an insider’s view
        Join us for this conversation with Dr. Clarance Lee, Senior Product Manger, about how Thermo Fisher is delivering the benefits of dPCR, along with high-throughput automation that saves time and enables diverse application areas. 
        31:47
      • Play video qPCR & dPCR Better Together - Studio of the Future
        qPCR & dPCR Better Together - Studio of the Future
        The Applied Biosystems QuantStudio real-time and digital PCR systems combined with Applied Biosystems Taqman chemistry deliver flexibility, precision and, confidence at every step.
        1:00
      • Play video Going Green with PCR to Detecting Grapevine Pathogens, with Lexi Heger| Absolute Gene-ius Podcast S4E1
        Going Green with PCR to Detecting Grapevine Pathogens, with Lexi Heger| Absolute Gene-ius Podcast S4E1
        In this episode of Absolute Gene-ius, Lexi Heger, PhD candidate at Michigan State University, explores how qPCR and digital PCR are transforming plant pathology.
        36:05
      • Play video ProFlex PCR Cinematic
        ProFlex PCR Cinematic
        Increase productivity with our PCR workflow solutions.
        0:34
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels