High performance PCR plastics for optimal PCR results
0:54
Description
Related Videos
"High-performance MicroAmp PCR and real-time PCR plastics for optimal PCR and qPCR results. Validated to work with Applied Biosystems thermal cyclers and real-time PCR instruments for over 25 years. MicroAmp plastics are designed for optimal heat transfer with thin-walled polypropylene wells and to reduce cross-contamination with raised well rims for effective sealing."