      • Stem Cell Research
      Three questions to ask when choosing a PCR enzyme
      1:17
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      Do you need to preserve sequence accuracy? If not, a standard DNA polymerase can be used to confirm presence or absence of target DNA. If, however, you will clone or perform sequencing, choose a high-fidelity proofreading polymerase for accurate sequence. Are you working with long DNA templates or looking to cut down cycling time? If so, choose a polymerase with a high processivity and extension rate. This will ensure efficient DNA synthesis and shorter reaction times. Are you encountering non-specific amplification? If you’re seeing too many bands that should not be there, choose a hot-start polymerase. This can help get more yield and sensitivity of the target.
      View More
      View Less
      Learn more about choosing PCR enzymes
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Related Videos
      In PCR Videos
      • Play video Absolute Gene-ius: S2 E8 Now that’s what I call a high-quality viral vector
        Absolute Gene-ius: S2 E8 Now that’s what I call a high-quality viral vector
        Dr. Cliff Froelich covers what vectors his team works with, what methods and technologies they use to monitor quality and safety, and he reminds us why it all matters.
        32:29
      • Play video Absolute Gene-ius: S2 E10 Automating accuracy – an insider’s view
        Absolute Gene-ius: S2 E10 Automating accuracy – an insider’s view
        Join us for this conversation with Dr. Clarance Lee, Senior Product Manger, about how Thermo Fisher is delivering the benefits of dPCR, along with high-throughput automation that saves time and enables diverse application areas. 
        31:47
      • Play video Kingfisher Apex Dx Cinematic
        Kingfisher Apex Dx Cinematic
        Thermo Scientific KingFisher instruments automate extraction of DNA, RNA, protein, and cells.
        1:05
      • Play video dPCR Academy - resDNA QA, QC
        dPCR Academy - resDNA QA, QC
        Webinar covering the principles of dPCR and its use in residual DNA detection for quality assurance and control.
        57:02
      • Play video Absolute Gene-ius: S2 E11 Somatic mosaic loss of sex chromosomes and kidney disease
        Absolute Gene-ius: S2 E11 Somatic mosaic loss of sex chromosomes and kidney disease
        Join us to hear about Dr. Parker Wilson's exciting work where he uses digital PCR and NGS methods to identify and quantify rare mutations associated with kidney disease.
        34:08
      • Play video Absolute Gene-ius: S2 E12 The Poisson perspective—counting out false positives
        Absolute Gene-ius: S2 E12 The Poisson perspective—counting out false positives
        We return to the topic of Poisson statistics and delve a bit deeper into how digital PCR (dPCR) handles topics like false positives or low-abundance and rare targets with Dave Bauer, PhD.
        17:46
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels