      • Stem Cell Research
      Boosting lab productivity with PCR enzymes
      1:00
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      Are you looking to boost your lab efficiency and productivity? Consider robust PCR enzymes that are designed for speed, simplicity, and accuracy. Speed: You can enjoy short PCR cycling time from fast DNA synthesis of Invitrogen Platinum II Taq Hot-Start DNA Polymerase at 15 sec/kb. Simplicity: You can shorten your PCR workflow by ~50% with Invitrogen Platinum Direct PCR Universal Master Mix. This product requires no DNA purification prior to PCR. Accuracy: You can spend less time troubleshooting downstream applications because amplicons of highly accurate sequences are generated by Invitrogen Platinum SuperFi II DNA Polymerase. This enzyme provides >300x higher fidelity than Taq DNA polymerase does.
      View More
      View Less
      Find your PCR boosters
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Related Videos
      In PCR Videos
      • Play video Love of dPCR - Fundamentals
        Love of dPCR - Fundamentals
        Webinar covering the principles of dPCR and its uses.
        47:42
      • Play video ProFlex PCR Cinematic
        ProFlex PCR Cinematic
        Increase productivity with our PCR workflow solutions.
        0:34
      • Play video Absolute Gene-ius: S2 E11 Somatic mosaic loss of sex chromosomes and kidney disease
        Absolute Gene-ius: S2 E11 Somatic mosaic loss of sex chromosomes and kidney disease
        Join us to hear about Dr. Parker Wilson's exciting work where he uses digital PCR and NGS methods to identify and quantify rare mutations associated with kidney disease.
        34:08
      • Play video Kingfisher Apex Dx Cinematic
        Kingfisher Apex Dx Cinematic
        Thermo Scientific KingFisher instruments automate extraction of DNA, RNA, protein, and cells.
        1:05
      • Play video qPCR & dPCR Better Together - Studio of the Future
        qPCR & dPCR Better Together - Studio of the Future
        The Applied Biosystems QuantStudio real-time and digital PCR systems combined with Applied Biosystems Taqman chemistry deliver flexibility, precision and, confidence at every step.
        1:00
      • Play video Absolute Gene-ius: S2 E12 The Poisson perspective—counting out false positives
        Absolute Gene-ius: S2 E12 The Poisson perspective—counting out false positives
        We return to the topic of Poisson statistics and delve a bit deeper into how digital PCR (dPCR) handles topics like false positives or low-abundance and rare targets with Dave Bauer, PhD.
        17:46
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels