      • Stem Cell Research
      Rare Mutation Detection with Digital PCR - ASHG 2014
      4:32
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      Product manager Dr. Iain Russell spoke with us about the new QuantStudio 3D rare mutation analysis solution. The solution includes a subset of wet lab–validated TaqMan SNP Genotyping Assays to detect and quantify the most common cancer-related mutations (e.g., EGFR, BRAF, KRAS, PIK3CA, and JAK2) and is designed using the full bioinformatics power of the TaqMan Assay design pipeline. Wet lab–validated specifically for the QuantStudio 3D, researchers are now able to quantify rare mutants at a prevalence as low as 0.1%.
      View More
      View Less
      Visit QuantStudio 3D Digital PCR System
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Related Videos
      In Real-Time PCR Videos
      • Play video VeriFlex™ Blocks Technology on Applied Biosystems™ QuantStudio RT-PCR Systems: Preview
        VeriFlex™ Blocks Technology on Applied Biosystems™ QuantStudio RT-PCR Systems: Preview
        Watch this short preview to learn how to optimize your PCR assays with VeriFlex Blocks technology
        0:33
      • Play video Enhance your qPCR experience with innovative and intuitive features: Preview
        Enhance your qPCR experience with innovative and intuitive features: Preview
        Watch this short preview to learn about the innovative features offered on Applied Biosystems™ QuantStudio RT-PCR Systems
        0:32
      • Play video Supply Chain & Trusted Supplier video – thermofisher.com/covid19EUA
        Supply Chain & Trusted Supplier video – thermofisher.com/covid19EUA
        Thermo Fisher Scientific is leading the world in COVID-19 testing. Our scale and depth of capabilities have never been more vital.
        3:05
      • Play video Supply Chain & Trusted Supplier video – thermofisher.com/covid19
        Supply Chain & Trusted Supplier video – thermofisher.com/covid19
        Thermo Fisher Scientific is leading the world in COVID-19 testing. Our scale and depth of capabilities have never been more vital.
        3:05
      • Play video Learn how multiplexing can help you become more efficient in the lab: Preview
        Learn how multiplexing can help you become more efficient in the lab: Preview
        Watch this short preview about how multiplexing, offered on Applied Biosystems™ QuantStudio RT-PCR Systems, can help you become more efficient.
        0:44
      • Play video PRRS 3.0 qPCR Preparing Template Demo
        PRRS 3.0 qPCR Preparing Template Demo
        The PRRS 3.0 kit uses qPCR technology to identify strains of Porcine Respiratory & Reproductive Syndrome Virus in pigs
        3:42
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels