Bitumen and asphalt testing - Dynamic Shear Rheometer (DSR) Thermo Scientific HAAKE MARS iQ Air Rheometer
4:12
The Thermo Scientific HAAKE MARS iQ Rheometer for dynamic shear testing according to AASHTO, ASTM, FGSV and DIN EN for bitumen and asphalt is ideal for routine test laboratories. The rheometer employs several smart functionalities to ensure precise and consistent results. Depending on the standard, it can be equipped with different measuring geometries and an appropriate temperature control. Changing between different configurations is easy, fast and failure-free with “Connect Assist” technology. You can initiate company specific SOPs via the convenient instrument touchscreen or with the rheometer software on the PC and always ensure, that the correct protocol is followed during the entire measurement. Some of the QC features of the HAAKE MARS iQ Rheometers and the HAAKE RheoWin Software are: 1. User guidance, with embedded individual videos and pictures to prevent handling failures 2. Automated data evaluation using your QC criteria to ensure correct result interpretation 3. Generation of reports according to specific company standards with automated data storage 4. Software offers real time display of graphical, numerical measuring data. Via touchscreen numerical real time data & measuring range bars can be monitored.