Invitrogen Bigfoot Cell Sorter - Time Savings and Higher Throughput
1:50
The Bigfoot Spectral Cell Sorter offers sort rates of >70,000 eps and analysis rates of >100,000 eps. The system is also capable of multi-way sorting into 1.5ml, 5ml, 15ml, and 50ml tubes with an easily configurable sort output tube holder. Using tubes, the sorter is capable of 6-way sorting and virtual 18-way sorting. Bigfoot can also perform 4-way sorting into 96 well plates, 8-way sorting into 384 well plates, and straight down sorting into 1536 well plates. The speed and recovery of the Bigfoot Spectral Cell Sorter is designed to exceed your expectations.