      • Stem Cell Research
      Introduction to protein degradation
      3:08
      • Share on Facebook
      • Tweet this video
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      Scientists have identified numerous proteins that are potential therapeutic targets. However, developing selective and effective therapeutics to target certain proteins has proven difficult due to their uninhabitable nature. Typically, small molecule drugs bind to functional sites on proteins. These sites are often conserved in enzyme families, which puts limits on the selectivity of drugs like kinase inhibitors. Targeted protein degradation has emerged as an advanced approach to reducing protein levels, and it can be selective for even the most difficult targets. The ubiquitin proteasome pathway (UPP) is a normal cellular process responsible for degradation of defective or surplus proteins. The UPP, involves 3 enzymes, E1, E2 and E3 ligase that mark the target protein with a polyubiquitin chain, directing them to the proteasome for degradation. Targeted protein degradation can help us understand the scientific value of targeted proteins. An example targeting the UPP is use of proteolysis targeting chimeras [2]. A proteolysis-targeting chimera can bind to both a protein of interest and a specific ubiquitin ligase, often E3 ligase, and bring them into close proximity. Successful complexation leads to ubiquitination and subsequent degradation of the protein. Proteolysis-targeting chimeras have been developed to study difficult targets, such as BRD4 and CDK9. Modulating the UPP can help reveal the effects of disease-related protein removal for difficult-to-target proteins and protein variants within minutes or hours, and it can be done in a reversible manner. Thermo Fisher Scientific has built the Protein Degradation Resource Center, an educational site that houses information about protein degradation pathways, assays, and assay reagents to support you in discovering your next therapeutic target.
      View More
      View Less
      Visit resource center
      Share
      • Share on Facebook
      • Tweet this video
      • Share on LinkedIn
      • Share via Email
      Related Videos
      In Recently Published Videos
      • Play video Box to bench & setup: Thermo Scientificâ„¢ TDE Series ultra-low temperature freezers
        Box to bench & setup: Thermo Scientificâ„¢ TDE Series ultra-low temperature freezers
        Unpacking and installing a new ultra-low temperature freezer can seem like a daunting task. In this video we discuss simple steps to making sure your installation goes as smoothly as possible.
        4:30
      • Play video Care & maintenance: How to minimize costly service calls and unnecessary down time
        Care & maintenance: How to minimize costly service calls and unnecessary down time
        Proper ULT freezer operation starts with understanding your equipment. In this video we discuss ways to help avoid unnecessary freezer alarms and downtime, including: load practices, facility electrical/circuit setup, door opening
        4:22
      • Play video Liquid biopsy analysis at Hospital Valdecilla using the QuantStudio Absolute Q dPCR System
        Liquid biopsy analysis at Hospital Valdecilla using the QuantStudio Absolute Q dPCR System
        Customer testimonial about liquid biopsy analysis at Hospital Valdecilla using the QuantStudio Absolute Q dPCR System
        2:50
      • Play video Liquid biopsy analysis at Hospital Puerta de Hierro using the QuantStudio Absolute Q dPCR System
        Liquid biopsy analysis at Hospital Puerta de Hierro using the QuantStudio Absolute Q dPCR System
        Customer testimonial about liquid biopsy analysis at Hospital Puerta de Hierro using the QuantStudio Absolute Q dPCR System
        2:10
      • Play video How to choose? What are technical data sheets and how can they help you?
        How to choose? What are technical data sheets and how can they help you?
        Technical data sheets provide information about electrical requirements, physical construction, operating conditions, and much more. In this video we discuss how to use this information to select and prepare for your ultra-low temperature freezer.
        5:08
      • Play video Box to bench & setup: Setting alarm limits on Thermo Scientificâ„¢ TSX ultra-low temperature freezers
        Box to bench & setup: Setting alarm limits on Thermo Scientificâ„¢ TSX ultra-low temperature freezers
        Setting the proper warm and cold alarm points on your ULT freezer is critical to maintaining sample security. In this video we will discuss best practices for assigning these alarm limits and how to set them on your TSX ULT freezer.
        2:06
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels