Absolute Gene-ius: S2 E12 The Poisson perspective—counting out false positives
17:46
Description
Related Videos
The statistics of Poisson distributions can seem complex at first but are simpler than you think, which is important to know given their relevance to dPCR. In short, they dictate the confidence you can have in the absolute quantification provided by dPCR.