      • Stem Cell Research
      Collibri 3' mRNA Library Prep for Illumina Systems
      2:56
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      Low cost gene expression studies are now possible from as few as 2 million reads in a single-end, 75 bp sequencing run on Illumina NGS systems. The Invitrogen Collibri 3’ mRNA Library Prep Kits for Illumina Systems use a blocking mechanism to capture only the 3’ end of each transcript, enabling shorter sequencing runs. Normalization (RPKM) is not required with this sequencing method because the transcript is degraded during library preparation, ensuring that gene expression results match sample complexity in the sequencing data. Globin-blocking oligos are included in the Invitrogen Collibri 3’ Blood mRNA Kit to enrich for gene expression by reducing the presence of globin in sequencing data. Collibri 3’ mRNA kits are compatible with all samples types and quality, including FFPE. Collibri 3’ Blood mRNA kits are compatible with all human samples.
      View More
      View Less
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Related Videos
      In Sequencing Videos
      • Play video Automated NGS with Genexus System
        Automated NGS with Genexus System
        Explore the automate NGS workflow, including purification from sample, sequencing, and analysis. Want more details? thermofisher.com/genexus
        3:41
      • Play video Dr. Sadikovic shares his experience using the Oncomine Myeloid assay for myeloid malignancies
        Dr. Sadikovic shares his experience using the Oncomine Myeloid assay for myeloid malignancies
        Hear how the Oncomine Myeloid Research assay allows for the assessment of a range of relevant variants for myeloid malignancies in a single assay
        1:59
      • Play video Genexus Dx Integrated Sequencer: Fast, Easy, & Automated CE-IVD NGS System
        Genexus Dx Integrated Sequencer: Fast, Easy, & Automated CE-IVD NGS System
        The CE-IVD marked Ion Torrent Genexus Dx Integrated Sequencer enables fast, easy, and automated clinical NGS.
        33:31
      • Play video IVF Cinematic
        IVF Cinematic
        Thermo Scientific Nunc in vitro fertilization (IVF) products are CE-marked Class II* for IVD devices and produced under ISO 13485 guidelines.
        1:59
      • Play video Collibri Stranded RNA Library Prep Kits for Illumina Systems
        Collibri Stranded RNA Library Prep Kits for Illumina Systems
        Collibri Stranded RNA Library Prep Kits for Illumina Systems
        2:25
      • Play video Case Studies of MSI Analysis & KRAS Variant Detection in Colorectal Cancer
        Case Studies of MSI Analysis & KRAS Variant Detection in Colorectal Cancer
        Webinar discussing the landscape of combination cancer therapies and how genotypic biomarkers data such as MSI analysis and KRAS sequencing are used.
        54:11
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels