Passaging reagents for PSC culture
3:35
Description
Related Videos
Dissocation of pluripotent stem cell (PSCs) can be achieved by different methods, including the use of either enzymatic or non-enzymatic reagents. Different workflows, including routine passaging, gene editing and stem cell differentiation, require varying degrees of dissociation. These varying degress of dissociation, from larger clumps to clusters of 2-3 cells or single cells, can be accomplished using different passaging reagents. Watch video to learn more about each >