      • Stem Cell Research
      Fast, Flexible, and Spectrally Brilliant: Invitrogen Attune Xenith Flow Cytometer
      1:00
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      Fast, flexible, and spectrally brilliant—simplify high-speed, high-parameter analysis with the Invitrogen Attune Xenith Flow Cytometer, powered by our legacy core acoustic hydrodynamic focusing technology. Experience remarkable speed, operating five times faster than most spectral cytometers, and benefit from innovative flexibility with seamless transitions between spectral unmixing and conventional flow cytometry on the same instrument using the same sample. Designed to handle challenging samples without clogging, the Attune Xenith Flow Cytometer boasts an advanced optical design featuring 6 lasers, 51 fluorescence detectors, and 6 scatter channels. Enjoy simplified maintenance with an intuitive touchscreen and advanced remote access diagnostics, including onboard cameras to ensure uninterrupted performance. Learn how the Invitrogen Attune Xenith Flow Cytometer can transform your research with speed, flexibility, and unparalleled performance. Visit thermofisher.com/attunexenith for more information.
      View More
      View Less
      Watch now
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Related Videos
      In Cell & Gene Therapy Videos
      • Play video Absolute Gene-ius S2 E1
        Absolute Gene-ius S2 E1
        A couple of reproductive biology experts
        0:28
      • Play video Automated Image Analysis with the Attune CytPix flow cytometer
        Automated Image Analysis with the Attune CytPix flow cytometer
        The Invitrogen™ Attune™ CytPix™ Flow Cytometer enables image capture and processing with the new version of the Invitrogen™ Attune™Cytometric Software. You can now measure cell morphology parameters concurrently with flow cytometric parameters.
        1:15
      • Play video Gibco™ CTS™ Compleo™ Fill and Finish System Fluid Path
        Gibco™ CTS™ Compleo™ Fill and Finish System Fluid Path
        This video showcases a fluid path for the CTS Compleo System for formulation and controlled filling into output containers for cell therapy manufacturing.
        2:47
      • Play video Absolute Gene-ius: S2 E6 What’s your vector, Victor?
        Absolute Gene-ius: S2 E6 What’s your vector, Victor?
        Curious about cell and gene therapy and the viral vectors used in these applications? Join us for a conversation to shed light on these topics.
        35:28
      • Play video Absolute Gene-ius: S2 E2 CAR-T loads of immunology insights
        Absolute Gene-ius: S2 E2 CAR-T loads of immunology insights
        This episode, which focuses on CAR-T cell therapy. Join us to meet Raquel Munoz and learn about what she thinks will be the future of cancer research
        31:59
      • Play video Gibco™ CTS™ Compleo™ Fill and Finish System Sampling
        Gibco™ CTS™ Compleo™ Fill and Finish System Sampling
        This video showcases one method of aseptic sampling performed using the Gibco CTS Compleo Fill and Finish System for cell therapy manufacturing.
        1:25
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels