Boosting lab productivity with PCR enzymes
1:00
Are you looking to boost your lab efficiency and productivity? Consider robust PCR enzymes that are designed for speed, simplicity, and accuracy. Speed: You can enjoy short PCR cycling time from fast DNA synthesis of Invitrogen Platinum II Taq Hot-Start DNA Polymerase at 15 sec/kb. Simplicity: You can shorten your PCR workflow by ~50% with Invitrogen Platinum Direct PCR Universal Master Mix. This product requires no DNA purification prior to PCR. Accuracy: You can spend less time troubleshooting downstream applications because amplicons of highly accurate sequences are generated by Invitrogen Platinum SuperFi II DNA Polymerase. This enzyme provides >300x higher fidelity than Taq DNA polymerase does.