      • Stem Cell Research
      How to perform a western wet tank transfer using the Invitrogen Mini Blot Module
      4:05
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Description
      Related Videos
      Once the proteins have been separated in the gel, they now can be transferred to a solid support membrane to start the western blotting workflow. In this video, you’ll learn how to perform a western transfer using the Mini Blot module in a Mini Gel Tank. Make sure the cassette clamps are removed before you set up your transfers. One gel is transferred per module, and two modules fit in one tank. Included with the Mini Blot Module kit is a roller, sponge pads, tweezers, and a quick reference card. Approximately 250 milliliters of Transfer Buffer is required for each transfer. Prepare Transfer Buffer by adding 25 milliliters of 20X Bolt Transfer Buffer, 50 milliliters of methanol, and 500 microliter of Bolt Antioxidant, and bring the total volume up to 500 milliliters with deionized water. Soak two sponge pads in transfer Buffer and Squeeze the pads to remove air bubbles. Open the gel cassette using a gel knife and carefully remove the wells and the foot of the gel, so that the entire gel is of equal thickness. Place a shallow dish on the bench and pour 1X transfer buffer to dip the filter papers into during sandwich assembly. Assemble the western transfer sandwich by placing the cathode or negative core on a flat surface. Pour 5 milliliters of transfer buffer into the core, then place a pre-soaked sponge pad on the cathode core followed by a pre-wetted filter paper. Next, use the blot roller to squeeze out the air bubbles from between each layer. Place the gel on the stack, again using the roller to remove air bubbles. Using tweezers, place the membrane on top of the gel and roll once more. Complete the stack by layering another pre-wetted filter paper on top of the membrane, removing bubbles, and placing pre-wetted sponge pads on top of the filter paper. Complete the assembly by placing the anode core on top of the sandwich and pressing the two module halves together. Insert the blot module into the Mini Gel Tank with the cathode core facing the front. The blot module should be seated with the electrodes contacting the electrode bar. Extra transfer buffer can be added to the module core to submerge the sandwich, but don’t fill it above the gasket. Add transfer Buffer or deionized water—about 225 milliliters to the chamber, just below the electrode bar. Make sure the power supply is off. Place the cover on the tank and plug the power leads into the power supply. Turn the power supply on to begin the transfer. For Nitrocellulose membranes, transfer at a constant 10 volts for 60 minutes. For PVDF membranes, transfer at 20 volts for 60 minutes. After the transfer is complete, turn off the power supply, disconnect the cords, open the lid by placing your thumb on the tank handles pulling the lid up with your fingers. Remove the module, pour out the buffer, and disassemble the module. Remove the membrane using tweezers. You are now ready to probe your membrane for western analysis. Find out more information about western blotting with the Mini Blot Module at thermofisher.com/westerntransfer
      View More
      View Less
      Learn more
      Share
      • Share on Facebook
      • Share on X
      • Share on LinkedIn
      • Share via Email
      Related Videos
      In Protein Biology Videos
      • Play video Advancing multiplex imaging for spatial proteomics research
        Advancing multiplex imaging for spatial proteomics research
        Learn about the Spatial Biology workflow at a high level and then understand how to image mIF with the EVOS S1000 Spatial Imaging System
        25:31
      • Play video Enhanced detection of low abundance proteins or dilute samples?
        Enhanced detection of low abundance proteins or dilute samples?
        Double your sample loading volume and boost the sensitivity of chemiluminescent detection.
        0:27
      • Play video Optilite Cinematic
        Optilite Cinematic
        Transform your diagnostic testing with the Optilite® Analyser.
        2:14
      • Play video Nature Conference: Spatial Biology 2024 | Advancing multiplex imaging for spatial proteomics research
        Nature Conference: Spatial Biology 2024 | Advancing multiplex imaging for spatial proteomics research
        Watch and learn about the advancements in multiplex imaging for spatial protein research, including the Invitrogen EVOS S1000 Spatial Imaging System, Invitrogen Aluora Spatial Amplification Reagents, primary antibodies conjugated to fluorophores
        25:31
      • Play video Be The Spark podcast: How is multiplex immunoassay changing the game in your research?
        Be The Spark podcast: How is multiplex immunoassay changing the game in your research?
        Welcome to "Be The Spark," the podcast where we delve into immunology and the cutting-edge science driving the field. In this episode, we discuss the advantage of multiplex bead-based immunoassays for cytokine profiling with Malia Edwards, Associate.
        11:07
      • Play video Run high performance protein gels in Bio-Rad’s Tank
        Run high performance protein gels in Bio-Rad’s Tank
        Get sharp, straight bands no matter your protein molecular weight, in as few as 14 minutes.
        0:31
      •  
      •  
      •  
      •  
      •  
      Brands
      • Thermo Scientific
      • Applied Biosystems
      • Invitrogen
      • Fisher Scientific
      • Unity Lab Services
      Shopping Tool
      • Product Selection Guides - US
      • Quick Order - US
      • Redeem a Quote
        * Please enter a valid quote
      • New Products
      • Promotions
      • Mobile & Desktop Apps
      • Shared Lists
      eSolutions
      • eProcurement
      • Supply Center
      • Instrument Management
      Support
      • Order Support
      • Training
      • Webinars
      • Blog
      • Social Media
      • Contact Us
      • Report a Site Issue
      Thermo Fisher Scientific
      • About Us
      • Careers
      • Investors
      • News
      • Responsibility
      • Trademarks
      United States
      • Terms & Conditions
      • Privacy Information Center
      • Price & Freight Policy
      Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.
      Ok
      Share this video
      Embed
      Size: x pixels